tert-Butyl-4-[(5-methyl-2-nitrophenyl)amino]piperidine-1-carboxylate ≥95%

Supplier: Matrix Scientific

056458-500MG 056458-1G
101910-578EA 278.46 USD
101910-578 101910-580
tert-Butyl-4-[(5-methyl-2-nitrophenyl)amino]piperidine-1-carboxylate ≥95%
tert-Butyl-4-[(5-methyl-2-nitrophenyl)amino]piperidine-1-carboxylate

Hazard: Irritant

Formula: C₁₇H₂₅N₃O₄
MW: 335.41 g/mol
Melting Pt: 102…105 °C
MDL Number: MFCD16037242
CAS Number: 950772-97-1

Order Now

Specification Test Results

Melting point 102-105°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR