tert-Butyl-6-hydroxy-1,2,3,4-tetrahydroisoquinoline-2-carboxylate ≥97%
Supplier: Matrix Scientific
Synonyms:
tert-Butyl 6-hydroxy-3,4-dihydroisoquinoline-2(1H)-carboxylate
056457-500MG
056457-1G
101910-574EA
98.46
USD
101910-574
101910-576
tert-Butyl-6-hydroxy-1,2,3,4-tetrahydroisoquinoline-2-carboxylate ≥97%
tert-Butyl-6-hydroxy-1,2,3,4-tetrahydroisoquinoline-2-carboxylate
Hazard: Irritant
Formula:
C₁₄H₁₉NO₃ MW: 249.31 g/mol Melting Pt: 99…100 °C |
MDL Number:
MFCD08275034 CAS Number: 158984-83-9 |
Specification Test Results
Melting point | 99-100°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...