tert-Butyl-6-cyano-1,2,3,4-tetrahydroisoquinoline-2-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: tert-Butyl 6-cyano-3,4-dihydroisoquinoline-2(1H)-carboxylate

056454-500MG 056454-1G
101910-562EA 692.31 USD
101910-562 101910-564
tert-Butyl-6-cyano-1,2,3,4-tetrahydroisoquinoline-2-carboxylate ≥95%
tert-Butyl-6-cyano-1,2,3,4-tetrahydroisoquinoline-2-carboxylate

Hazard: Irritant

Formula: C₁₅H₁₈N₂O₂
MW: 258.32 g/mol
Melting Pt: 144…146 °C
MDL Number: MFCD11043755
CAS Number: 166398-33-0

Order Now

Specification Test Results

Melting point 144-146°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR