tert-Butyl-4-[3-(benzylamino)-2-cyanophenyl]piperazine-1-carboxylate ≥95%

Supplier: Matrix Scientific

056452-500MG 056452-1G
101910-554EA 556.92 USD
101910-554 101910-556
tert-Butyl-4-[3-(benzylamino)-2-cyanophenyl]piperazine-1-carboxylate ≥95%
tert-Butyl-4-[3-(benzylamino)-2-cyanophenyl]piperazine-1-carboxylate

Hazard: Irritant

Formula: C₂₃H₂₈N₄O₂
MW: 392.51 g/mol
Melting Pt: 166…168 °C
MDL Number: MFCD16710298
CAS Number: 1242268-05-8

Order Now

Specification Test Results

Melting point 166-168°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR