tert-Butyl-3-amino-8-thia-4,6,11-triazatricyclo[7.4.0.0
Supplier: Matrix Scientific
056451-500MG
056451-1G
101910-550EA
375.38
USD
101910-550
101910-552
tert-Butyl-3-amino-8-thia-4,6,11-triazatricyclo[7.4.0.0
tert-Butyl-3-amino-8-thia-4,6,11-triazatricyclo[7.4.0.0^{2,7}]trideca-1(9),2(7),3,5-tetraene-11-carboxylate
Hazard: Irritant
Formula:
C₁₄H₁₈N₄O₂S MW: 306.39 g/mol Melting Pt: 194…196 °C |
MDL Number:
MFCD16710285 CAS Number: 1196798-60-3 |
Specification Test Results
Melting point | 194-196°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...