1H-Pyrrolo[2,3-b]pyridin-3-ylmethanol ≥95%

Supplier: Matrix Scientific

Synonyms: (1H-Pyrrolo[2,3-b]pyridin-3-yl)methanol

056444-500MG 056444-1G
101910-526EA 523.08 USD
101910-526 101910-528
1H-Pyrrolo[2,3-b]pyridin-3-ylmethanol ≥95%
1H-Pyrrolo[2,3-b]pyridin-3-ylmethanol

Hazard: Irritant

Formula: C₈H₈N₂O
MW: 148.17 g/mol
Melting Pt: 171…173 °C
MDL Number: MFCD11054203
CAS Number: 1065100-83-5

Order Now

Specification Test Results

Melting point 171-173°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR