(1-Methyl-1H-[1,2,4]triazol-3-yl)methanol ≥95%

Supplier: Matrix Scientific

056439-500MG 056439-1G 056439-5G
101910-504EA 127.69 USD
101910-504 101910-506 101910-508
(1-Methyl-1H-[1,2,4]triazol-3-yl)methanol ≥95%
(1-Methyl-1H-[1,2,4]triazol-3-yl)methanol

Hazard: Irritant

Formula: C₄H₇N₃O
MW: 113.1200 g/mol
Melting Pt: 72…75 °C
MDL Number: MFCD16710296
CAS Number: 135242-93-2

Order Now

Specification Test Results

Melting point 72-75°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR