Methyl-6-oxo-5H,6H-pyrrolo[1,2-a]quinoxaline-3-carboxylate ≥95%

Supplier: Matrix Scientific

056433-500MG 056433-1G
101910-484EA 556.92 USD
101910-484 101910-486
Methyl-6-oxo-5H,6H-pyrrolo[1,2-a]quinoxaline-3-carboxylate ≥95%
Methyl-6-oxo-5H,6H-pyrrolo[1,2-a]quinoxaline-3-carboxylate

Hazard: Irritant

Formula: C₁₃H₁₀N₂O₃
Melting Pt: 256…257 °C (dec.)
MDL Number: MFCD16710293
CAS Number: 1006385-87-0

Order Now

Specification Test Results

Melting point 256-257°C(d


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR