Methyl-4,5-dihydrospiro[pyrrolo(1,2-a)quinoxaline-4,4'-piperidine]-7-carboxylate hydrochloride ≥95%

Supplier: Matrix Scientific

056429-500MG 056429-1G
101910-472EA 556.92 USD
101910-472 101910-474
Methyl-4,5-dihydrospiro[pyrrolo(1,2-a)quinoxaline-4,4'-piperidine]-7-carboxylate hydrochloride ≥95%
Methyl-4,5-dihydrospiro[pyrrolo(1,2-a)quinoxaline-4,4'-piperidine]-7-carboxylate hydrochloride

Hazard: Irritant

Formula: C₁₇H₂₀ClN₃O₂
MW: 333.82 g/mol
Melting Pt: 278…279 °C (dec.)
MDL Number: MFCD16710291
CAS Number: 1242268-06-9

Order Now

Specification Test Results

Melting point 278-279°C(d


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR