7-Methyl-4,5-dihydrospiro[pyrrolo(1,2-a)quinoxaline-4,4'-piperidine]hydrochloride ≥95%

Supplier: Matrix Scientific

056428-500MG 056428-1G
101910-468EA 556.92 USD
101910-468 101910-470
7-Methyl-4,5-dihydrospiro[pyrrolo(1,2-a)quinoxaline-4,4'-piperidine]hydrochloride ≥95%
7-Methyl-4,5-dihydrospiro[pyrrolo(1,2-a)quinoxaline-4,4'-piperidine]hydrochloride

Hazard: Irritant

Formula: C₁₆H₂₀ClN₃
MW: 289.81 g/mol
Melting Pt: 300…301 °C (dec.)
MDL Number: MFCD16710290
CAS Number: 1242268-31-0

Order Now

Specification Test Results

Melting point 300-301°C(d

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR