Methyl-4-amino-3-(ethylamino)benzoate ≥95%

Supplier: Matrix Scientific

056422-500MG 056422-1G
101910-448EA 556.92 USD
101910-448 101910-450
Methyl-4-amino-3-(ethylamino)benzoate ≥95%
Methyl-4-amino-3-(ethylamino)benzoate

Hazard: Irritant

Formula: C₁₀H₁₄N₂O₂
Melting Pt: 94…96 °C
MDL Number: MFCD16710286
CAS Number: 1242268-09-2

Order Now

Specification Test Results

Melting point 94-96°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR