3-Methoxy-5H,6H-pyrrolo[1,2-a]quinoxalin-6-one ≥95%

Supplier: Matrix Scientific

056418-500MG 056418-1G
101910-434EA 556.92 USD
101910-434 101910-436
3-Methoxy-5H,6H-pyrrolo[1,2-a]quinoxalin-6-one ≥95%
3-Methoxy-5H,6H-pyrrolo[1,2-a]quinoxalin-6-one

Hazard: Irritant

Formula: C₁₂H₁₀N₂O₂
Melting Pt: 249…251 °C
MDL Number: MFCD16710282
CAS Number: 160657-07-8

Order Now

Specification Test Results

Melting point 249-251°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR