N-{Imidazo[1,2-a]pyridin-5-yl}-2,2-dimethylpropanamide ≥95%

Supplier: Matrix Scientific

056410-500MG 056410-1G
101910-396EA 278.46 USD
101910-396 101910-398
N-{Imidazo[1,2-a]pyridin-5-yl}-2,2-dimethylpropanamide ≥95%
N-{Imidazo[1,2-a]pyridin-5-yl}-2,2-dimethylpropanamide

Hazard: Irritant

Formula: C₁₂H₁₅N₃O
Melting Pt: 126…129 °C
MDL Number: MFCD16710271
CAS Number: 1242268-22-9

Order Now

Specification Test Results

Melting point 126-129°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR