Ethyl-4-cyano-1H-pyrazole-3-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Ethyl 4-cyano-1H-pyrazole-5-carboxylate

056400-500MG 056400-1G
101910-354EA 556.92 USD
101910-354 101910-356
Ethyl-4-cyano-1H-pyrazole-3-carboxylate ≥95%
Ethyl-4-cyano-1H-pyrazole-3-carboxylate

Hazard: Irritant

Formula: C₇H₇N₃O₂
MW: 165.15 g/mol
Melting Pt: 223…225 °C
MDL Number: MFCD00664125
CAS Number: 318497-88-0

Order Now

Specification Test Results

Melting point 223-225°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR