3,4-Dihydro-2H-1,4-benzoxazin-6-amine hydrochloride ≥95%

Supplier: Matrix Scientific

Synonyms: 3,4-Dihydro-2H-benzo[b][1,4]oxazin-6-amine hydrochloride

056393-500MG 056393-1G
101910-326EA 423.08 USD
101910-326 101910-328
3,4-Dihydro-2H-1,4-benzoxazin-6-amine hydrochloride ≥95%
3,4-Dihydro-2H-1,4-benzoxazin-6-amine hydrochloride

Hazard: Irritant

Formula: C₈H₁₁ClN₂O
MW: 186.64 g/mol
Melting Pt: >250 °C
MDL Number: MFCD16710277
CAS Number: 207596-59-6

Order Now

Specification Test Results

Melting point >250°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR