(3-Chloropyridin-2-yl)methanol ≥95%

Supplier: Matrix Scientific

056385-500MG 056385-1G
101910-296EA 269.23 USD
101910-296 101910-298
(3-Chloropyridin-2-yl)methanol ≥95%
(3-Chloropyridin-2-yl)methanol

Hazard: Irritant

Formula: C₆H₆ClNO
Melting Pt: 42…44 °C (lit.)
MDL Number: MFCD11110226
CAS Number: 60588-81-0

Order Now

Specification Test Results

Melting point 42-44°C(lit


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR