4-Chloro-N-methylpicolinamide ≥95%
Supplier: Matrix Scientific
056381-500MG
056381-1G
056381-5G
101910-278EA
556.92
USD
101910-278
101910-280
101910-282
4-Chloro-N-methylpicolinamide ≥95%
4-Chloro-N-methylpicolinamide
Hazard: Irritant/Store in Freezer
Formula:
C₇H₇ClN₂O Melting Pt: 40…44 °C |
MDL Number:
MFCD02185921 CAS Number: 220000-87-3 |
Specification Test Results
Melting point | 40-44°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...