3-(Chloromethyl)-1-methyl-1H-1,2,4-triazole hydrochloride ≥95%

Supplier: Matrix Scientific

056380-500MG 056380-1G 056380-5G
101910-272EA 326.15 USD
101910-272 101910-274 101910-276
3-(Chloromethyl)-1-methyl-1H-1,2,4-triazole hydrochloride ≥95%
3-(Chloromethyl)-1-methyl-1H-1,2,4-triazole hydrochloride

Hazard: Irritant

Formula: C₄H₇Cl₂N₃
Melting Pt: 69…70 °C
MDL Number: MFCD09701417
CAS Number: 135206-76-7

Order Now

Specification Test Results

Melting point 69-70°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR