6-Chloro-5-iodopyridine-3-carbonitrile ≥95%

Supplier: Matrix Scientific

Synonyms: 6-Chloro-5-iodonicotinonitrile

056377-500MG 056377-1G
101910-260EA 623.08 USD
101910-260 101910-262
6-Chloro-5-iodopyridine-3-carbonitrile ≥95%
6-Chloro-5-iodopyridine-3-carbonitrile

Hazard: Irritant

Formula: C₆H₂ClIN₂
MW: 264.45 g/mol
Melting Pt: 150…152 °C
MDL Number: MFCD16710295
CAS Number: 856213-95-1

Order Now

Specification Test Results

Melting point 150-152°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR