4-{3-Chloro-5-[(4-cyanophenyl)amino]phenoxy}-3,5-dimethylbenzonitrile ≥95%

Supplier: Matrix Scientific

056375-500MG 056375-1G
101910-252EA 375.38 USD
101910-252 101910-254
4-{3-Chloro-5-[(4-cyanophenyl)amino]phenoxy}-3,5-dimethylbenzonitrile ≥95%
4-{3-Chloro-5-[(4-cyanophenyl)amino]phenoxy}-3,5-dimethylbenzonitrile

Hazard: Irritant

Formula: C₂₂H₁₆ClN₃O
Melting Pt: 278…280 °C
MDL Number: MFCD16710267
CAS Number: 1242268-27-4

Order Now

Specification Test Results

Melting point 278-280°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR