6-Bromo-3-methyl-2,3-dihydro-1,3-benzoxazol-2-one ≥95%

Supplier: Matrix Scientific

056372-500MG 056372-1G
101910-240EA 200 USD
101910-240 101910-242
6-Bromo-3-methyl-2,3-dihydro-1,3-benzoxazol-2-one ≥95%
6-Bromo-3-methyl-2,3-dihydro-1,3-benzoxazol-2-one

Hazard: Irritant

Formula: C₈H₆BrNO₂
Melting Pt: 146…147 °C
MDL Number: MFCD00694789
CAS Number: 67927-44-0

Order Now

Specification Test Results

Melting point 146-147°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR