2-(Benzylamino)-6-fluorobenzonitrile ≥95%

Supplier: Matrix Scientific

056370-500MG 056370-1G
101910-232EA 278.46 USD
101910-232 101910-234
2-(Benzylamino)-6-fluorobenzonitrile ≥95%
2-(Benzylamino)-6-fluorobenzonitrile

Hazard: Irritant

Formula: C₁₄H₁₁FN₂
Melting Pt: 120…122 °C
MDL Number: MFCD12427793
CAS Number: 1155987-86-2

Order Now

Specification Test Results

Melting point 120-122°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR