1,2,3,4-Tetrahydrobenzo[b]azepin-5-one ≥95%

Supplier: Matrix Scientific

Synonyms: 1,2,3,4-Tetrahydro-benzo[b]azepin-5-one, 1,2,3,4-Tetrahydrobenzazepine-5-one, 2,3,4,5-tetrahydro-1H-1-benzazepin-5-one

055503-1G 055503-5G
101909-552EA 300 USD
101909-552 101909-554
1,2,3,4-Tetrahydrobenzo[b]azepin-5-one ≥95%
1,2,3,4-Tetrahydrobenzo[b]azepin-5-one

Hazard: Irritant

Formula: C₁₀H₁₁NO
MW: 161.204 g/mol
Melting Pt: 70…72 °C
MDL Number: MFCD03426404
CAS Number: 1127-74-8

Order Now

Specification Test Results

Melting point 70-72°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR