(2S,4R)-1-(tert-Butoxycarbonyl)-4-hydroxyhexahydro-2-pyridine carboxylic acid ≥95%

Supplier: Matrix Scientific

Synonyms: (2S,4R)-4-Hydroxypiperidine-2-carboxylic acid, N-BOC protected, (2S,4R)-1-(tert-Butoxycarbonyl)-4-hydroxypiperidine-2-carboxylic acid

055500-500MG 055500-1G
101909-542EA 921.54 USD
101909-542 101909-544
(2S,4R)-1-(tert-Butoxycarbonyl)-4-hydroxyhexahydro-2-pyridine carboxylic acid ≥95%
(2S,4R)-1-(tert-Butoxycarbonyl)-4-hydroxyhexahydro-2-pyridine carboxylic acid

Hazard: Irritant

Formula: C₁₁H₁₉NO₅
MW: 245.28 g/mol
Melting Pt: >163 °C (dec.)
MDL Number: MFCD04112667
CAS Number: 955016-25-8

Order Now

Specification Test Results

Melting point >163°C(dec)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR