3-Nitro-4-(1H-pyrrol-1-yl)benzenol ≥95%

Supplier: Matrix Scientific

055496-500MG 055496-1G
101909-528EA 278.46 USD
101909-528 101909-530
3-Nitro-4-(1H-pyrrol-1-yl)benzenol ≥95%
3-Nitro-4-(1H-pyrrol-1-yl)benzenol

Hazard: Irritant

Formula: C₁₀H₈N₂O₃
Melting Pt: 156 °C
MDL Number: MFCD16622793
CAS Number: 251649-40-8

Order Now

Specification Test Results

Melting point 156°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR