N-(6-Nitro-1,3-benzodioxol-5-yl)acetamide ≥95%

Supplier: Matrix Scientific

055495-500MG 055495-1G
101909-524EA 375.38 USD
101909-524 101909-526
N-(6-Nitro-1,3-benzodioxol-5-yl)acetamide ≥95%
N-(6-Nitro-1,3-benzodioxol-5-yl)acetamide

Hazard: Irritant

Formula: C₉H₈N₂O₅
Melting Pt: 209…211 °C
MDL Number: MFCD00188397
CAS Number: 81864-14-4

Order Now

Specification Test Results

Melting point 209-211°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR