6-(Methylsulfonyl)nicotinonitrile ≥97%

Supplier: Matrix Scientific

055494-500MG 055494-1G
101909-520EA 200 USD
101909-520 101909-522
6-(Methylsulfonyl)nicotinonitrile ≥97%
6-(Methylsulfonyl)nicotinonitrile

Hazard: Irritant

Formula: C₇H₆N₂O₂S
MW: 182.2 g/mol
Melting Pt: 134…136 °C
MDL Number: MFCD16622798
CAS Number: 66154-68-5

Order Now

Specification Test Results

Melting point 134-136°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR