Methyl-2-(methylamino)-5-nitronicotinate ≥95%

Supplier: Matrix Scientific

055493-500MG 055493-1G
101909-516EA 375.38 USD
101909-516 101909-518
Methyl-2-(methylamino)-5-nitronicotinate ≥95%
Methyl-2-(methylamino)-5-nitronicotinate

Hazard: Irritant

Formula: C₈H₉N₃O₄
MW: 211.18 g/mol
Melting Pt: 152…153 °C
MDL Number: MFCD16622796
CAS Number: 1228182-64-6

Order Now

Specification Test Results

Melting point 152-153°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR