Methyl-5-chloropicolinate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 5-chloropyridine-2-carboxylate

055488-500MG 055488-1G
101909-496EA 592.31 USD
101909-496 101909-498
Methyl-5-chloropicolinate ≥95%
Methyl 5-chloropicolinate

Hazard: Irritant

Formula: C₇H₆ClNO₂
MW: 171.5800 g/mol
Melting Pt: 85-88°C
MDL Number: MFCD09702466
CAS Number: 132308-19-1

Order Now

Specification Test Results

Melting point 80-82°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR