Urocanic acid ≥95%

Supplier: Matrix Scientific

Synonyms: Imidazole-4-acrylic acid, 4-Imidazoleacrylic acid, 3-(1H-Imidazol-4-yl)acrylic acid

055485-500MG 055485-1G
101909-486EA 181.54 USD
101909-486 101909-488
Urocanic acid ≥95%
Urocanic acid

Hazard: Irritant

Formula: C₆H₆N₂O₂
MW: 138.126 g/mol
Melting Pt: >215 °C (dec.)
MDL Number: MFCD00005203
CAS Number: 104-98-3

Order Now

Specification Test Results

Melting point >215°C(dec)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR