2-Hydroxy-3-methoxy-3,3-diphenylpropanoic acid ≥95%

Supplier: Matrix Scientific

055484-500MG 055484-1G 055484-5G
101909-480EA 40 USD
101909-480 101909-482 101909-484
2-Hydroxy-3-methoxy-3,3-diphenylpropanoic acid ≥95%
2-Hydroxy-3-methoxy-3,3-diphenylpropanoic acid

Hazard: Irritant

Formula: C₁₆H₁₆O₄
Melting Pt: >95 °C (dec.)
MDL Number: MFCD13152279
CAS Number: 178306-51-9

Order Now

Specification Test Results

Melting point >95°C(dec)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR