2-[(E)-(4-Chlorophenyl)methylidene]-1-hydrazinecarboximidamide ≥95%

Supplier: Matrix Scientific

055475-500MG 055475-1G 055475-5G
101909-446EA 278.46 USD
101909-446 101909-448 101909-450
2-[(E)-(4-Chlorophenyl)methylidene]-1-hydrazinecarboximidamide ≥95%
2-[(E)-(4-Chlorophenyl)methylidene]-1-hydrazinecarboximidamide

Hazard: Irritant

Formula: C₈H₉ClN₄
Melting Pt: 195…197 °C
MDL Number: MFCD00205397
CAS Number: 13308-88-8

Order Now

Specification Test Results

Melting point 195-197°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR