4-Chloro-2-iodo-6-methylaniline ≥95%

Supplier: Matrix Scientific

055474-500MG 055474-1G 055474-5G
101909-440EA 121.54 USD
101909-440 101909-442 101909-444
4-Chloro-2-iodo-6-methylaniline ≥95%
4-Chloro-2-iodo-6-methylaniline

Hazard: Irritant

Formula: C₇H₇CllN
Melting Pt: 47…49 °C
MDL Number: MFCD16622764
CAS Number: 69705-29-9

Order Now

Specification Test Results

Melting point 47-49°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR