5-Chloro-1-benzothiophene-2-carbonitrile ≥95%

Supplier: Matrix Scientific

055472-500MG 055472-1G
101909-432EA 1109.23 USD
101909-432 101909-434
5-Chloro-1-benzothiophene-2-carbonitrile ≥95%
5-Chloro-1-benzothiophene-2-carbonitrile

Hazard: Irritant

Formula: C₉H₄ClNS
Melting Pt: 130…132 °C
MDL Number: MFCD13181028
CAS Number: 23622-24-4

Order Now

Specification Test Results

Melting point 130-132°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR