5-Chloro-1-benzothiophene-2-carbonyl chloride ≥95%

Supplier: Matrix Scientific

055471-500MG 055471-1G
101909-428EA 747.69 USD
101909-428 101909-430
5-Chloro-1-benzothiophene-2-carbonyl chloride ≥95%
5-Chloro-1-benzothiophene-2-carbonyl chloride

Hazard: Irritant

Formula: C₉H₄Cl₂OS
Melting Pt: 98…100 °C
MDL Number: MFCD16622799
CAS Number: 13771-97-6

Order Now

Specification Test Results

Melting point 98-100°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR