1-Benzhydryl-3-phenylazetane ≥95%

Supplier: Matrix Scientific

Synonyms: 1-Benzhydryl-3-phenylazetidine

055466-500MG 055466-1G 055466-5G
101909-404EA 173.85 USD
101909-404 101909-406 101909-408
1-Benzhydryl-3-phenylazetane ≥95%
1-Benzhydryl-3-phenylazetane

Hazard: Irritant

Formula: C₂₂H₂₁N
MW: 299.42 g/mol
Melting Pt: 77…78 °C
MDL Number: MFCD16622779
CAS Number: 913814-30-9

Order Now

Specification Test Results

Melting point 77-78°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR