3-Amino-4-(1H-pyrrol-1-yl)benzenol ≥95%

Supplier: Matrix Scientific

055465-500MG 055465-1G
101909-400EA 556.92 USD
101909-400 101909-402
3-Amino-4-(1H-pyrrol-1-yl)benzenol ≥95%
3-Amino-4-(1H-pyrrol-1-yl)benzenol

Hazard: Irritant

Formula: C₁₀H₁₀N₂O
Melting Pt: 147…149 °C
MDL Number: MFCD16622801
CAS Number: 1228182-66-8

Order Now

Specification Test Results

Melting point 147-149°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR