tert-Butyl-N-[1-(4-chloro-2-formylphenyl)-3-piperidinyl]carbamate ≥95%

Supplier: Matrix Scientific

055459-500MG 055459-1G
101909-378EA 375.38 USD
101909-378 101909-380
tert-Butyl-N-[1-(4-chloro-2-formylphenyl)-3-piperidinyl]carbamate ≥95%
tert-Butyl-N-[1-(4-chloro-2-formylphenyl)-3-piperidinyl]carbamate

Hazard: Irritant

Formula: C₁₇H₂₃ClN₂O₃
Melting Pt: 111…113 °C
MDL Number: MFCD16622783
CAS Number: 1228183-71-8

Order Now

Specification Test Results

Melting point 111-113°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR