6-(tert-Butoxycarbonyl)-6-azaspiro[2.5]octane-1-carboxylic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 6-Azaspiro[2.5]octane-1-carboxylic acid, N-BOC protected

055455-500MG 055455-1G
101909-364EA 230.77 USD
101909-364 101909-366
6-(tert-Butoxycarbonyl)-6-azaspiro[2.5]octane-1-carboxylic acid ≥95%
6-(tert-Butoxycarbonyl)-6-azaspiro[2.5]octane-1-carboxylic acid

Hazard: Irritant

Formula: C₁₃H₂₁NO₄
Melting Pt: 128…129 °C
MDL Number: MFCD11007805
CAS Number: 871727-05-8

Order Now

Specification Test Results

Melting point 128-129°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR