2-(Methylsulfonyl)nicotinonitrile ≥95%

Supplier: Matrix Scientific

055452-500MG 055452-1G
101909-354EA 278.46 USD
101909-354 101909-356
2-(Methylsulfonyl)nicotinonitrile ≥95%
2-(Methylsulfonyl)nicotinonitrile

Hazard: Irritant

Formula: C₇H₆N₂O₂S
Melting Pt: 103…105 °C
MDL Number: MFCD16622788
CAS Number: 66154-66-3

Order Now

Specification Test Results

Melting point 103-105°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR