Methyl-2-hydroxy-5-nitronicotinate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 5-nitro-2-oxo-1,2-dihydropyridine-3-carboxylate

055450-500MG 055450-1G 055450-5G
101909-344EA 278.46 USD
101909-344 101909-346 101909-348
Methyl-2-hydroxy-5-nitronicotinate ≥95%
Methyl-2-hydroxy-5-nitronicotinate

Hazard: Irritant

Formula: C₇H₆N₂O₅
MW: 198.14 g/mol
Melting Pt: 161…164 °C
MDL Number: MFCD07783889
CAS Number: 856579-28-7

Order Now

Specification Test Results

Melting point 161-164°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR