7-Methoxy-4,5-dihydrospiro[pyrrolo(1,2-a)quinoxaline-4,4'-piperidine] hydrochloride ≥95%

Supplier: Matrix Scientific

055447-500MG 055447-1G 055447-5G
101909-326EA 747.69 USD
101909-326 101909-328 101909-330
7-Methoxy-4,5-dihydrospiro[pyrrolo(1,2-a)quinoxaline-4,4'-piperidine] hydrochloride ≥95%
7-Methoxy-4,5-dihydrospiro[pyrrolo(1,2-a)quinoxaline-4,4'-piperidine] hydrochloride

Hazard: Irritant

Formula: C₁₆H₂₀ClN₃O
Melting Pt: 267…268 °C
MDL Number: MFCD16622786
CAS Number: 1228182-68-0

Order Now

Specification Test Results

Melting point 267-268°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR