2-Hydroxy-5-nitronicotinic acid ≥97%

Supplier: Matrix Scientific

Synonyms: 5-Nitro-2-oxo-3-pyridinecarboxylic Acid

055446-1G 055446-5G
101909-322EA 158.46 USD
101909-322 101909-324
2-Hydroxy-5-nitronicotinic acid ≥97%
2-Hydroxy-5-nitronicotinic acid

Hazard: Irritant

Formula: C₆H₄N₂O₅
MW: 184.11 g/mol
Melting Pt: 265°C
MDL Number: MFCD05669267
CAS Number: 6854-07-5

Order Now

Specification Test Results

Melting point 245-247°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR