6-[cis-2,6-Dimethylmorpholin-4-yl]pyridin-3-amine dihydrochloride ≥95%

Supplier: Matrix Scientific

055443-500MG 055443-1G 055443-5G
101909-308EA 200 USD
101909-308 101909-310 101909-312
6-[cis-2,6-Dimethylmorpholin-4-yl]pyridin-3-amine dihydrochloride ≥95%
6-[cis-2,6-Dimethylmorpholin-4-yl]pyridin-3-amine dihydrochloride

Hazard: Irritant

Formula: C₁₁H₁₉Cl₂N₃O
MW: 280.2 g/mol
Melting Pt: 237…239 °C
MDL Number: MFCD16622770
CAS Number: 1282295-12-8

Order Now

Specification Test Results

Melting point 237-239°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR