5-Chloro-2-hydrazinylpyridine-4-carboxylic acid hydrochloride ≥95%

Supplier: Matrix Scientific

055438-500MG 055438-1G
101909-288EA 556.92 USD
101909-288 101909-290
5-Chloro-2-hydrazinylpyridine-4-carboxylic acid hydrochloride ≥95%
5-Chloro-2-hydrazinylpyridine-4-carboxylic acid hydrochloride

Hazard: Irritant

Formula: C₆H₇Cl₂N₃O₂
Melting Pt: 176…178 °C
MDL Number: MFCD16622784
CAS Number: 1228182-73-7

Order Now

Specification Test Results

Melting point 176-178°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR