5-Chloro-1-benzothiophene-2-carbaldehyde ≥95%

Supplier: Matrix Scientific

Synonyms: 5-Chlorobenzo[b]thiophene-2-carbaldehyde

055437-500MG 055437-1G
101909-284EA 921.54 USD
101909-284 101909-286
5-Chloro-1-benzothiophene-2-carbaldehyde ≥95%
5-Chloro-1-benzothiophene-2-carbaldehyde

Hazard: Irritant

Formula: C₉H₅ClOS
MW: 196.66 g/mol
Melting Pt: 132…134 °C
MDL Number: MFCD16622780
CAS Number: 28540-51-4

Order Now

Specification Test Results

Melting point 132-134°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR