(5-Chloro-1-benzothiophen-2-yl)methanol ≥95%

Supplier: Matrix Scientific

055436-500MG 055436-1G
101909-280EA 529.23 USD
101909-280 101909-282
(5-Chloro-1-benzothiophen-2-yl)methanol ≥95%
(5-Chloro-1-benzothiophen-2-yl)methanol

Hazard: Irritant

Formula: C₉H₇ClOS
Melting Pt: 123…125 °C
MDL Number: MFCD12032278
CAS Number: 13771-71-6

Order Now

Specification Test Results

Melting point 123-125°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR