2-(Bromomethyl)-5-chloro-1-benzothiophene ≥95%

Supplier: Matrix Scientific

055433-500MG 055433-1G
101909-268EA 800 USD
101909-268 101909-270
2-(Bromomethyl)-5-chloro-1-benzothiophene ≥95%
2-(Bromomethyl)-5-chloro-1-benzothiophene

Hazard: Irritant

Formula: C₉H₆BrClS
Melting Pt: 82…84 °C
MDL Number: MFCD16622781
CAS Number: 99592-53-7

Order Now

Specification Test Results

Melting point 82-84°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR