4-Amino-5-iodo-2-methoxybenzenecarboxylic acid ≥95%
Supplier: Matrix Scientific
Synonyms:
4-Amino-5-iodo-2-methoxybenzoic acid
055430-500MG
055430-1G
055430-5G
101909-252EA
409.23
USD
101909-252
101909-254
101909-256
4-Amino-5-iodo-2-methoxybenzenecarboxylic acid ≥95%
4-Amino-5-iodo-2-methoxybenzenecarboxylic acid
Hazard: Irritant
Formula:
C₈H₈INO₃ MW: 293.06 g/mol Melting Pt: 185…187 °C |
MDL Number:
MFCD16622772 CAS Number: 155928-39-5 |
Specification Test Results
Melting point | 185-187°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...