N-Allyl-N'-(2-cyanophenyl)urea ≥95%

Supplier: Matrix Scientific

055428-500MG 055428-1G 055428-5G
101909-242EA 181.54 USD
101909-242 101909-244 101909-246
N-Allyl-N'-(2-cyanophenyl)urea ≥95%
N-Allyl-N'-(2-cyanophenyl)urea

Hazard: Irritant

Formula: C₁₁H₁₁N₃O
Melting Pt: 173…175 °C
MDL Number: MFCD16622763
CAS Number: 122372-27-4

Order Now

Specification Test Results

Melting point 173-175°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR